Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rom-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047549.4 | NP_001041014.1 | 1 - 2 | 273 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |