Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047236.4 | NP_001040701.2 | 2 - 3 | 2232 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |