Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003249.1 | NP_001003249.1 | 3 - 4 | 473 | 113 | |
GenBank mRNA | L76257.1 | - | 3 - 4 | 608 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |