Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003333.1 | NP_001003333.1 | 273 | 71 | ||
GenBank mRNA | AB004268.1 | - | 239 | 71 | ||
D17397.1 | - | 273 | 71 | |||
HG965175.1 | - | 187 | 71 | |||
HG965176.1 | - | 187 | 71 | |||
HG965177.1 | - | 187 | 71 | |||
HG965178.1 | - | 187 | 71 | |||
HG965179.1 | - | 187 | 71 | |||
HG965180.1 | - | 187 | 71 | |||
HG965181.1 | - | 187 | 71 | |||
HG965182.1 | - | 187 | 71 | |||
HG965183.1 | - | 187 | 71 | |||
HG965184.1 | - | 187 | 71 | |||
HG965185.1 | - | 187 | 71 | |||
HG965186.1 | - | 187 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |