Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002976.2 | NP_001002976.1 | 3 - 4 | 363 | 92 | |
XM_022404314.1 | XP_022260022.1 | 4 - 5 | 805 | 92 | ||
GenBank mRNA | AB101519.1 | - | 3 - 4 | 363 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |