Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002948.2 | NP_001002948.1 | 4 - 5 | 512 | 137 | |
XM_022407411.1 | XP_022263119.1 | 4 - 5 | 713 | 137 | ||
GenBank mRNA | AB125102.1 | - | 4 - 5 | 513 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |