Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121722.2 | XP_013977197.1 | 2 | 2918 | 66 | |
XM_014121724.2 | XP_013977199.1 | 9 | 3761 | 66 | ||
GenBank mRNA | AY137582.1 | - | 1 | 1061 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |