Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018035.1 | NP_001018045.1 | 7 - 8 | 1927 | 69 | |
XM_005628839.3 | XP_005628896.1 | 7 - 8 | 2215 | 69 | ||
XM_005628841.3 | XP_005628898.1 | 6 - 7 | 1985 | 69 | ||
XM_005628842.3 | XP_005628899.1 | 6 - 7 | 1748 | 69 | ||
XM_014119231.2 | XP_013974706.1 | 7 - 8 | 2136 | 69 | ||
XM_022427665.1 | XP_022283373.1 | 7 - 8 | 1926 | 69 | ||
XM_022427666.1 | XP_022283374.1 | 7 - 8 | 1923 | 69 | ||
XM_022427667.1 | XP_022283375.1 | 6 - 7 | 1922 | 69 | ||
XM_022427668.1 | XP_022283376.1 | 6 - 7 | 1761 | 69 | ||
GenBank mRNA | DQ008575.1 | - | 7 - 8 | 1927 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |