Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003313.1 | NP_001003313.1 | 4 - 5 | 465 | 81 | |
XM_022415927.1 | XP_022271635.1 | 3 - 4 | 402 | 81 | ||
GenBank mRNA | J05466.1 | - | 4 - 5 | 465 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |