Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313810.1 | NP_001300739.1 | 2 - 3 | 280 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |