Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080725.1 | NP_001074194.1 | 6 - 7 | 1577 | 94 | |
XM_005619513.3 | XP_005619570.1 | 7 - 8 | 3872 | 94 | ||
XM_005619514.3 | XP_005619571.1 | 7 - 8 | 4971 | 94 | ||
XM_022418028.1 | XP_022273736.1 | 7 - 8 | 3934 | 94 | ||
XM_022418029.1 | XP_022273737.1 | 7 - 8 | 3784 | 94 | ||
GenBank mRNA | EF102104.1 | - | 6 - 7 | 1577 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |