Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022412897.1 | XP_022268605.1 | 4 - 5 | 610 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |