Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014377.1 | NP_001014399.1 | 3 - 4 | 436 | 80 | |
XM_022425718.1 | XP_022281426.1 | 4 - 5 | 590 | 80 | ||
XM_022425719.1 | XP_022281427.1 | 4 - 5 | 708 | 80 | ||
XM_022425720.1 | XP_022281428.1 | 4 - 5 | 590 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Inventoried Probe spans exons |