Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425120.1 | XP_022280828.1 | 253 | 57 | ||
XM_022425121.1 | XP_022280829.1 | 258 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris |