Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313834.1 | NP_001300763.1 | 3 - 4 | 615 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |