Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630568.2 | XP_005630625.1 | 4 - 5 | 3629 | 57 | |
XM_005630569.2 | XP_005630626.1 | 4 - 5 | 3313 | 57 | ||
XM_022404829.1 | XP_022260537.1 | 4 - 5 | 3284 | 57 | ||
XM_022404830.1 | XP_022260538.1 | 2 - 3 | 2017 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |