Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426887.1 | XP_022282595.1 | 2 - 3 | 619 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |