Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313837.1 | NP_001300766.1 | 13 - 14 | 1028 | 73 | |
XM_022421052.1 | XP_022276760.1 | 14 - 15 | 1122 | 73 | ||
XM_022421090.1 | XP_022276798.1 | 13 - 14 | 1077 | 73 | ||
XM_022421113.1 | XP_022276821.1 | 14 - 15 | 1171 | 73 | ||
XM_022421139.1 | XP_022276847.1 | 10 - 11 | 558 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |