Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547012.5 | XP_547012.4 | 418 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |