Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845089.5 | XP_850182.1 | 3 - 4 | 333 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |