Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405409.1 | XP_022261117.1 | 6 - 7 | 1400 | 81 | |
XM_022405410.1 | XP_022261118.1 | 5 - 6 | 1265 | 81 | ||
XM_540610.6 | XP_540610.2 | 5 - 6 | 1229 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |