Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640051.3 | XP_005640108.1 | 3 - 4 | 450 | 88 | |
XM_847554.5 | XP_852647.1 | 3 - 4 | 450 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |