Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423233.1 | XP_022278941.1 | 2 - 3 | 714 | 67 | |
XM_022423234.1 | XP_022278942.1 | 2 - 3 | 718 | 67 | ||
XM_022423235.1 | XP_022278943.1 | 3 - 4 | 648 | 67 | ||
XM_022423236.1 | XP_022278944.1 | 3 - 4 | 892 | 67 | ||
XM_022423237.1 | XP_022278945.1 | 2 - 3 | 333 | 67 | ||
XM_847459.5 | XP_852552.1 | 2 - 3 | 470 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |