Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARHSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434858.3 | XP_003434906.1 | 2 - 3 | 318 | 71 | |
XM_022420081.1 | XP_022275789.1 | 2 - 3 | 275 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |