Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634904.3 | XP_005634961.1 | 2 - 3 | 474 | 101 | |
XM_542943.5 | XP_542943.2 | 2 - 3 | 348 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |