Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618894.3 | XP_005618951.1 | 5 | 1481 | 109 | |
XM_005618897.3 | XP_005618954.1 | 5 | 1469 | 109 | ||
XM_022417436.1 | XP_022273144.1 | 5 | 1470 | 109 | ||
XM_022417437.1 | XP_022273145.1 | 5 | 1498 | 109 | ||
XM_022417438.1 | XP_022273146.1 | 5 | 1444 | 109 | ||
XM_022417439.1 | XP_022273147.1 | 4 | 1386 | 109 | ||
XM_546110.5 | XP_546110.1 | 5 | 1374 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Amplicon spans exons and probe does not span exons |