Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633881.3 | XP_005633938.1 | 1 | 757 | 160 | |
XM_005633883.3 | XP_005633940.1 | 1 | 721 | 160 | ||
XM_843276.5 | XP_848369.1 | 1 | 804 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |