Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629823.3 | XP_005629880.1 | 309 | 96 | ||
XM_022403995.1 | XP_022259703.1 | 378 | 96 | |||
XM_022403996.1 | XP_022259704.1 | 227 | 96 | |||
XM_022403997.1 | XP_022259705.1 | 613 | 96 | |||
XM_022403998.1 | XP_022259706.1 | 376 | 96 | |||
XM_022403999.1 | XP_022259707.1 | 377 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |