Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417591.1 | XP_022273299.1 | 1 | 120 | 154 | |
XM_859212.4 | XP_864305.1 | 2 | 410 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Amplicon spans exons and probe does not span exons |