Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620554.3 | XP_005620611.1 | 7678 | 133 | ||
XM_005620555.3 | XP_005620612.1 | 7712 | 133 | |||
XM_005620556.3 | XP_005620613.1 | 7672 | 133 | |||
XM_005620557.3 | XP_005620614.1 | 7677 | 133 | |||
XM_022419078.1 | XP_022274786.1 | 7709 | 133 | |||
XM_022419079.1 | XP_022274787.1 | 7772 | 133 | |||
XM_022419080.1 | XP_022274788.1 | 7769 | 133 | |||
XM_546778.5 | XP_546778.2 | 7672 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |