Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639887.3 | XP_005639944.1 | 7 | 1169 | 98 | |
XM_005639888.3 | XP_005639945.1 | 8 | 1324 | 98 | ||
XM_022414322.1 | XP_022270030.1 | 8 | 1409 | 98 | ||
XM_022414323.1 | XP_022270031.1 | 9 | 1503 | 98 | ||
XM_535853.6 | XP_535853.3 | 7 | 1227 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |