Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547420.6 | XP_547420.2 | 1231 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |