Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251946.1 | NP_001238875.1 | 3 - 4 | 336 | 114 | |
GenBank mRNA | DN267006.1 | - | 3 - 4 | 336 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |