Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxacb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498525.2 | XP_003498573.2 | 2 - 3 | 1982 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |