Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513122.1 | XP_003513170.1 | 3 - 4 | 564 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |