Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652396.1 | XP_007650586.1 | 2 - 3 | 644 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |