Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497863.2 | XP_003497911.1 | 2 - 3 | 854 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |