Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500122.1 | XP_003500170.1 | 3 - 4 | 482 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |