Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptplb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640866.1 | XP_007639056.1 | 5 - 6 | 632 | 127 | |
GenBank mRNA | JI884823.1 | - | 5 - 6 | 632 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Probe spans exons |