Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498835.2 | XP_003498883.2 | 1 - 2 | 364 | 79 | |
GenBank mRNA | JI886370.1 | - | 1 - 2 | 364 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |