Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511799.1 | XP_003511847.1 | 1 | 2926 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |