Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645443.1 | XP_007643633.1 | 5 - 6 | 652 | 108 | |
XM_007645444.1 | XP_007643634.1 | 5 - 6 | 637 | 108 | ||
GenBank mRNA | JI863066.1 | - | 4 - 5 | 598 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |