Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511048.2 | XP_003511096.1 | 2 - 3 | 1878 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Recommended Assay Probe spans exons |