Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498499.1 | XP_003498547.1 | 2 - 3 | 334 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |