Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640389.1 | XP_007638579.1 | 2 - 3 | 6693 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |