Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497003.2 | XP_003497051.1 | 1 - 2 | 154 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |