Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503493.2 | XP_003503541.1 | 3 - 4 | 2066 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |