Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496083.2 | XP_003496131.1 | 5 - 6 | 753 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |