Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501267.1 | XP_003501315.1 | 1 - 2 | 899 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Probe spans exons |