Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510181.2 | XP_003510229.1 | 2 - 3 | 134 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |